Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP5IF1
Entrez Gene ID
716870
Gene Name
ATP synthase inhibitory factor subunit 1
Gene Aliases
ATPIF1, EGK_00460
Chromosome Location
Chr.1: 196484756 - 196486713 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260663.1 NP_001247592.1 - 101 85
GenBank mRNA JU472972.1 - - 101 85
JV563352.1 - - 177 85
JV633820.1 - - 172 85

View Full Product Details