Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC12
Entrez Gene ID
707758
Gene Name
trafficking protein particle complex 12
Gene Aliases
TTC15
Chromosome Location
Chr.13: 105313693 - 105417791 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111612.2 XP_014967098.2 - 1493 71
XM_015111613.2 XP_014967099.2 - 1454 71
XM_015111616.2 XP_014967102.2 - 1493 71
XM_028832011.1 XP_028687844.1 - 1559 71
XM_028832012.1 XP_028687845.1 - 1493 71
GenBank mRNA JU325657.1 - - 906 71
JV546840.1 - - 921 71

View Full Product Details