Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCER1G
Entrez Gene ID
720291
Gene Name
Fc fragment of IgE receptor Ig
Gene Aliases
-
Chromosome Location
Chr.1: 89731037 - 89734869 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265840.2 NP_001252769.1 - 179 89
GenBank mRNA JU321435.1 - - 173 89
JU472311.1 - - 166 89
JV045676.1 - - 179 89

View Full Product Details