Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
718599
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
EGK_01431
Chromosome Location
Chr.1: 94228502 - 94237057 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257481.1 NP_001244410.1 - 982 69
XM_015110067.2 XP_014965553.1 - 473 69
XM_015110071.2 XP_014965557.1 - 428 69
XM_015110079.1 XP_014965565.1 - 1126 69
XM_028845997.1 XP_028701830.1 - 700 69
XM_028846002.1 XP_028701835.1 - 1022 69
XM_028846007.1 XP_028701840.1 - 1000 69
GenBank mRNA JU321339.1 - - 982 69
JU322927.1 - - 982 69
JU471886.1 - - 965 69
JV045616.1 - - 980 69

View Full Product Details