Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA50
Entrez Gene ID
709764
Gene Name
N-alpha-acetyltransferase 50, NatE catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.2: 162511203 - 162538312 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261371.2 NP_001248300.1 - 209 134
GenBank mRNA JU336491.1 - - 312 134
JU471806.1 - - 218 134
JV043838.1 - - 211 134
JV637198.1 - - 312 134
JV697188.1 - - 314 134

View Full Product Details