Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC11A
Entrez Gene ID
719381
Gene Name
C-type lectin domain containing 11A
Gene Aliases
-
Chromosome Location
Chr.19: 50629812 - 50637451 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001116136.4 XP_001116136.3 - 1152 114
XM_028839724.1 XP_028695557.1 - 532 114
GenBank mRNA JV635682.1 - - 678 114

View Full Product Details