Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE3
Entrez Gene ID
711390
Gene Name
jade family PHD finger 3
Gene Aliases
PHF16
Chromosome Location
Chr.X: 46874904 - 47025469 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100171.4 XP_001100171.3 - 1036 79
XM_002806209.3 XP_002806255.2 - 1276 79
XM_028842510.1 XP_028698343.1 - 1173 79
XM_028842511.1 XP_028698344.1 - 1147 79
GenBank mRNA JU337313.1 - - 1048 79
JU478015.1 - - 978 79
JV048826.1 - - 1131 79
JV666670.1 - - 1134 79

View Full Product Details