Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD2BP2
Entrez Gene ID
709961
Gene Name
CD2 cytoplasmic tail binding protein 2
Gene Aliases
EGK_12679
Chromosome Location
Chr.20: 27747430 - 27751935 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260696.1 NP_001247625.1 - 1017 75
GenBank mRNA JU332503.1 - - 1014 75
JU473976.1 - - 1004 75
JV045903.1 - - 1017 75
JV491995.1 - - 1003 75

View Full Product Details