Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GMPPB
- Entrez Gene ID
-
705454
- Gene Name
- GDP-mannose pyrophosphorylase B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 105837424 - 105839890 on Build Mmul_10
- Gene Symbol
- RNF123
- Entrez Gene ID
-
705340
- Gene Name
- ring finger protein 123
- Gene Aliases
-
EGK_11630
- Chromosome Location
-
Chr.2: 105803401 - 105837454 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU346973.1 |
- |
- |
4773 |
105 |
JU476736.1 |
- |
- |
5314 |
105 |
JU476737.1 |
- |
- |
5233 |
105 |
View Full Product Details