Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO5A
Entrez Gene ID
574273
Gene Name
myosin VA
Gene Aliases
MYOA
Chromosome Location
Chr.7: 28683735 - 28902808 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015142265.2 XP_014997751.1 - 1160 132
XM_015142266.2 XP_014997752.1 - 1119 132
XM_015142267.2 XP_014997753.1 - 1276 132
XM_015142268.2 XP_014997754.1 - 1119 132
XM_015142269.2 XP_014997755.1 - 1119 132
XM_015142270.2 XP_014997756.1 - 1276 132
XM_015142271.2 XP_014997757.1 - 1119 132
XM_028850935.1 XP_028706768.1 - 1119 132

View Full Product Details