Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU2
Entrez Gene ID
706065
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
EFHA1
Chromosome Location
Chr.17: 1671111 - 1777532 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001089388.4 XP_001089388.3 - 763 166
XM_015120728.2 XP_014976214.2 - 814 166
XM_015120729.2 XP_014976215.2 - 724 166
XM_028837518.1 XP_028693351.1 - 867 166
GenBank mRNA JU333393.1 - - 759 166
JV046451.1 - - 771 166

View Full Product Details