Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMAT2
Entrez Gene ID
697247
Gene Name
zinc finger matrin-type 2
Gene Aliases
-
Chromosome Location
Chr.6: 138081810 - 138088838 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261767.1 NP_001248696.1 - 122 80
GenBank mRNA JU327774.1 - - 85 80
JU474458.1 - - 122 80
JV489642.1 - - 132 80

View Full Product Details