Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
694759
Gene Name
solute carrier family 25 member 32
Gene Aliases
EGK_19190
Chromosome Location
Chr.8: 103677312 - 103692378 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266738.1 NP_001253667.1 - 628 146
GenBank mRNA JU321064.1 - - 580 146
JU472299.1 - - 462 146
JV044660.1 - - 628 146
JV637782.1 - - 613 146

View Full Product Details