Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB29
Entrez Gene ID
697184
Gene Name
RAB29, member RAS oncogene family
Gene Aliases
EGK_01737, RAB7L1
Chromosome Location
Chr.1: 63291028 - 63298815 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260953.1 NP_001247882.1 - 279 71
XM_015118289.2 XP_014973775.1 - 363 71
XM_015118296.2 XP_014973782.1 - 351 71
XM_015118304.2 XP_014973790.1 - 337 71
GenBank mRNA JU321140.1 - - 342 71
JU472082.1 - - 279 71
JV704065.1 - - 350 71

View Full Product Details