Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC8B
Entrez Gene ID
707348
Gene Name
kelch domain containing 8B
Gene Aliases
EGK_11644
Chromosome Location
Chr.2: 105245936 - 105256330 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257592.1 NP_001244521.1 - 927 92
XM_028843267.1 XP_028699100.1 - 1208 92
GenBank mRNA JU320493.1 - - 927 92
JV043989.1 - - 966 92
JV529168.1 - - 999 92
JV566819.1 - - 987 92

View Full Product Details