Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIANP
Entrez Gene ID
713548
Gene Name
PILR alpha associated neural protein
Gene Aliases
C11H12orf53, C12orf53, EGK_03284
Chromosome Location
Chr.11: 6852161 - 6861050 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194405.3 NP_001181334.1 - 249 93
XM_015150806.2 XP_015006292.1 - 181 93
XM_015150808.2 XP_015006294.1 - 287 93
XM_015150810.2 XP_015006296.1 - 287 93
XM_028828980.1 XP_028684813.1 - 162 93
XM_028828981.1 XP_028684814.1 - 182 93
GenBank mRNA JU333283.1 - - 96 93
JV535845.1 - - 99 93
JV667722.1 - - 96 93

View Full Product Details