Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPH3
Entrez Gene ID
694760
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
EGK_19802
Chromosome Location
Chr.9: 69582142 - 69593110 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266749.1 NP_001253678.1 - 831 107
XM_015147324.2 XP_015002810.1 - 890 107
XM_015147325.2 XP_015002811.1 - 955 107
XM_015147326.2 XP_015002812.1 - 845 107
XM_015147327.2 XP_015002813.1 - 826 107
XM_015147328.2 XP_015002814.1 - 910 107
XM_028853261.1 XP_028709094.1 - 751 107
XM_028853262.1 XP_028709095.1 - 732 107
XM_028853263.1 XP_028709096.1 - 816 107
XM_028853264.1 XP_028709097.1 - 706 107
XM_028853265.1 XP_028709098.1 - 687 107
GenBank mRNA JU320257.1 - - 850 107
JU334281.1 - - 786 107
JU334282.1 - - 831 107
JV649904.1 - - 1027 107

View Full Product Details