Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNAL1
Entrez Gene ID
712636
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
Chromosome Location
Chr.17: 9280398 - 9375424 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194119.3 NP_001181048.1 - 1249 148
XM_001099323.4 XP_001099323.1 - 1298 148
XM_015120818.2 XP_014976304.1 - 1315 148
XM_015120819.2 XP_014976305.1 - 1517 148
XM_015120820.2 XP_014976306.1 - 1217 148
GenBank mRNA JU336650.1 - - 1201 148
JU473391.1 - - 1227 148

View Full Product Details