Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
711343
Gene Name
craniofacial development protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 62273946 - 62424530 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194469.2 NP_001181398.1 - 229 88
GenBank mRNA JU321259.1 - - 195 88
JU472355.1 - - 196 88
JV044365.1 - - 189 88
JV637979.1 - - 205 88

View Full Product Details