Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP1
Entrez Gene ID
704448
Gene Name
STEAP family member 1
Gene Aliases
-
Chromosome Location
Chr.3: 115916123 - 115926744 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015134145.1 XP_014989631.1 - 1017 91
XM_028846095.1 XP_028701928.1 - 765 91
XM_028846096.1 XP_028701929.1 - 739 91
GenBank mRNA JV057891.1 - - 142 91
JV648263.1 - - 137 91

View Full Product Details