Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERPINH1
Entrez Gene ID
696171
Gene Name
serpin family H member 1
Gene Aliases
-
Chromosome Location
Chr.14: 68629463 - 68639949 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261087.1 NP_001248016.1 - 736 114
GenBank mRNA JU328560.1 - - 294 114
JU477636.1 - - 774 114
JV048570.1 - - 736 114
JV664397.1 - - 860 114

View Full Product Details