Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRACDL
Entrez Gene ID
712938
Gene Name
CRACD like
Gene Aliases
KIAA1211L
Chromosome Location
Chr.13: 2586459 - 2732754 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028832203.1 XP_028688036.1 - 1465 64
XM_028832204.1 XP_028688037.1 - 1031 64
XM_028832205.1 XP_028688038.1 - 1057 64
XM_028832206.1 XP_028688039.1 - 867 64
XM_028832207.1 XP_028688040.1 - 834 64
XM_028832208.1 XP_028688041.1 - 1034 64
XM_028832209.1 XP_028688042.1 - 1111 64

View Full Product Details