Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B9D1
Entrez Gene ID
702949
Gene Name
B9 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.16: 18829047 - 18852822 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266722.2 NP_001253651.1 - 629 61
GenBank mRNA JU333993.1 - - 629 61

View Full Product Details