Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPB
Entrez Gene ID
718461
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit B
Gene Aliases
EGK_06649
Chromosome Location
Chr.14: 65271590 - 65421145 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261302.1 NP_001248231.1 - 1619 108
XM_015115132.2 XP_014970618.1 - 1688 108
XM_015115133.2 XP_014970619.1 - 1689 108
XM_015115134.2 XP_014970620.1 - 1602 108
GenBank mRNA JU327232.1 - - 837 108
JU477357.1 - - 1619 108
JV724320.1 - - 844 108

View Full Product Details