Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEUROD6
Entrez Gene ID
694337
Gene Name
neuronal differentiation 6
Gene Aliases
EGK_13798
Chromosome Location
Chr.3: 83782718 - 83786368 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260574.1 NP_001247503.1 - 304 118
GenBank mRNA JU320533.1 - - 304 118

View Full Product Details