Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOSTDC1
Entrez Gene ID
709577
Gene Name
sclerostin domain containing 1
Gene Aliases
EGK_13889
Chromosome Location
Chr.3: 98471005 - 98544270 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194311.3 NP_001181240.1 - 766 62
GenBank mRNA JV577688.1 - - 274 62

View Full Product Details