Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNG
Entrez Gene ID
713769
Gene Name
taxilin gamma
Gene Aliases
CXorf15, EGK_20298, TXLNGX
Chromosome Location
Chr.X: 16475529 - 16534578 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127003.2 XP_014982489.2 - 743 84
XM_028842169.1 XP_028698002.1 - 743 84
GenBank mRNA JU335433.1 - - 678 84

View Full Product Details