Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMA1
Entrez Gene ID
716395
Gene Name
chymase 1
Gene Aliases
EGK_18076
Chromosome Location
Chr.7: 86583968 - 86586894 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194633.3 NP_001181562.2 - 621 68

View Full Product Details