Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC3
Entrez Gene ID
694884
Gene Name
coiled-coil domain containing 3
Gene Aliases
-
Chromosome Location
Chr.9: 14047110 - 14192658 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193755.2 NP_001180684.1 - 562 74
XM_028853267.1 XP_028709100.1 - 563 74
GenBank mRNA JU327176.1 - - 207 74
JV522287.1 - - 229 74
JV600232.1 - - 402 74
JV724660.1 - - 304 74
JV724661.1 - - 304 74

View Full Product Details