Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPPR1
Entrez Gene ID
714164
Gene Name
phospholipid phosphatase related 1
Gene Aliases
EGK_07491, LPPR1
Chromosome Location
Chr.15: 41577673 - 41870759 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266056.1 NP_001252985.1 - 1164 69
GenBank mRNA JU322099.1 - - 1164 69
JV549051.1 - - 790 69

View Full Product Details