Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF19B
Entrez Gene ID
710523
Gene Name
ring finger protein 19B
Gene Aliases
IBRDC3
Chromosome Location
Chr.1: 191672670 - 191707888 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015134625.2 XP_014990111.2 - 1729 94
XM_015134636.2 XP_014990122.2 - 1768 94
XM_015134643.2 XP_014990129.2 - 1763 94
XM_028836364.1 XP_028692197.1 - 1768 94
XM_028836369.1 XP_028692202.1 - 1775 94
XM_028836372.1 XP_028692205.1 - 1777 94

View Full Product Details