Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALU
Entrez Gene ID
699730
Gene Name
calumenin
Gene Aliases
-
Chromosome Location
Chr.3: 154984308 - 155017602 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260801.1 NP_001247730.1 - 308 71
GenBank mRNA JU320008.1 - - 308 71

View Full Product Details