Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP4
Entrez Gene ID
704483
Gene Name
phospholipid phosphatase 4
Gene Aliases
PPAPDC1, PPAPDC1A
Chromosome Location
Chr.9: 120945938 - 121091502 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266703.1 NP_001253632.1 - 389 79
XM_028853381.1 XP_028709214.1 - 344 79
GenBank mRNA DQ148090.1 - - 351 79
JU333277.1 - - 393 79
JV045160.1 - - 389 79

View Full Product Details