Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP4
Entrez Gene ID
693393
Gene Name
SH3 domain binding protein 4
Gene Aliases
-
Chromosome Location
Chr.12: 122364236 - 122468880 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261601.1 NP_001248530.1 - 331 121
XM_028830360.1 XP_028686193.1 - 524 121
XM_028830361.1 XP_028686194.1 - 439 121
XM_028830362.1 XP_028686195.1 - 574 121
XM_028830363.1 XP_028686196.1 - 373 121
XM_028830364.1 XP_028686197.1 - 822 121
GenBank mRNA JU321255.1 - - 423 121
JV043405.1 - - 331 121
JV638180.1 - - 425 121

View Full Product Details