Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRKB
Entrez Gene ID
707023
Gene Name
TP53RK binding protein
Gene Aliases
-
Chromosome Location
Chr.13: 35109959 - 35117768 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105330.4 XP_001105330.3 - 422 140
XM_015112657.2 XP_014968143.1 - 431 140
XM_015112660.2 XP_014968146.1 - 353 140
XM_015112661.2 XP_014968147.1 - 352 140
GenBank mRNA JU326371.1 - - 267 140
JV062462.1 - - 380 140

View Full Product Details