Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM248
Entrez Gene ID
695654
Gene Name
transmembrane protein 248
Gene Aliases
C3H7orf42, C7orf42
Chromosome Location
Chr.3: 53057692 - 53096054 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193777.2 NP_001180706.1 - 1041 105
GenBank mRNA JU320843.1 - - 1009 105
JU473650.1 - - 968 105
JV044150.1 - - 961 105

View Full Product Details