Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F6
Entrez Gene ID
697195
Gene Name
solute carrier family 35 member F6
Gene Aliases
C13H2orf18, C2orf18, EGK_05140
Chromosome Location
Chr.13: 82193286 - 82208829 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193806.1 NP_001180735.1 - 223 76
GenBank mRNA JU321250.1 - - 219 76
JU321251.1 - - 219 76
JU476167.1 - - 203 76
JU476168.1 - - 203 76
JV639346.1 - - 228 76

View Full Product Details