Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYAB
Entrez Gene ID
710747
Gene Name
crystallin alpha B
Gene Aliases
EGK_06886
Chromosome Location
Chr.14: 105055681 - 105070284 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260901.1 NP_001247830.1 - 237 77
XM_028832669.1 XP_028688502.1 - 630 77
XM_028832670.1 XP_028688503.1 - 435 77
XM_028832671.1 XP_028688504.1 - 591 77
GenBank mRNA JU470931.1 - - 236 77
JV044848.1 - - 237 77
JV568055.1 - - 232 77
JV635704.1 - - 256 77

View Full Product Details