Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM76A
Entrez Gene ID
716420
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
Chromosome Location
Chr.1: 196960157 - 196998280 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001111916.4 XP_001111916.1 - 239 72
XM_001111995.4 XP_001111995.1 - 238 72
GenBank mRNA JU320723.1 - - 173 72
JU320724.1 - - 173 72
JV044953.1 - - 152 72

View Full Product Details