Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR37
Entrez Gene ID
696628
Gene Name
G protein-coupled receptor 37
Gene Aliases
-
Chromosome Location
Chr.3: 151129744 - 151149613 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257869.1 NP_001244798.1 - 1676 73
GenBank mRNA JU334111.1 - - 1676 73

View Full Product Details