Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRM5
Entrez Gene ID
574330
Gene Name
cholinergic receptor muscarinic 5
Gene Aliases
EGK_17314
Chromosome Location
Chr.7: 10421727 - 10423325 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032931.1 NP_001028103.1 - 291 63
GenBank mRNA AF026264.1 - - 291 63
AF285237.1 - - 174 63

View Full Product Details