Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHA
Entrez Gene ID
574379
Gene Name
inhibin subunit alpha
Gene Aliases
EGK_04834
Chromosome Location
Chr.12: 107003332 - 107006615 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY574370.1 - - 94 107

View Full Product Details