Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMB5
Entrez Gene ID
677719
Gene Name
proteasome 20S subunit beta 5
Gene Aliases
EGK_18013
Chromosome Location
Chr.7: 85061812 - 85071287 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260845.2 NP_001247774.1 - 530 65
XM_015143267.1 XP_014998753.1 - 428 65
GenBank mRNA AY680474.1 - - 66 65
JU335537.1 - - 518 65
JU476465.1 - - 518 65
JV047706.1 - - 530 65

View Full Product Details