Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL44
Entrez Gene ID
677716
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
-
Chromosome Location
Chr.12: 111408997 - 111421968 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111171.2 XP_014966657.1 - 1114 69
GenBank mRNA AY680570.1 - - 270 69

View Full Product Details