Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMB6
Entrez Gene ID
710028
Gene Name
proteasome 20S subunit beta 6
Gene Aliases
EGK_08055
Chromosome Location
Chr.16: 4628201 - 4630550 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195714.1 NP_001182643.1 - 593 55
GenBank mRNA CN648676.1 - - 593 55
DQ148155.1 - - 138 55
JU327181.1 - - 514 55
JU489932.1 - - 571 55

View Full Product Details