Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACR3
Entrez Gene ID
653013
Gene Name
tachykinin receptor 3
Gene Aliases
EGK_15980, NK-3
Chromosome Location
Chr.5: 101996866 - 102101565 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037862.1 NP_001032951.1 - 891 73
GenBank mRNA AB180075.1 - - 891 73

View Full Product Details