Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKG2D
Entrez Gene ID
574240
Gene Name
NKG2D protein
Gene Aliases
KLRK1, NKG2-D
Chromosome Location
Chr.11: 9359059 - 9376449 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032889.1 NP_001028061.1 - 171 85
XM_015150990.2 XP_015006476.2 - 346 85
GenBank mRNA AF190943.1 - - 171 85
AJ554302.1 - - 158 85
JU474751.1 - - 299 85

View Full Product Details