Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKG2D
Entrez Gene ID
574240
Gene Name
NKG2D protein
Gene Aliases
KLRK1, NKG2-D
Chromosome Location
Chr.11: 9359059 - 9376449 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF190944.1 - - 202 143

View Full Product Details