Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCN6
Entrez Gene ID
574148
Gene Name
lipocalin 6
Gene Aliases
-
Chromosome Location
Chr.15: 1481365 - 1485860 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032831.1 NP_001028003.1 - 555 98
GenBank mRNA AF303085.1 - - 555 98

View Full Product Details