Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMP3
Entrez Gene ID
574213
Gene Name
lysosomal associated membrane protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 88379254 - 88434521 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032872.1 NP_001028044.1 - 1118 98
GenBank mRNA AF416334.1 - - 1118 98
JU484479.1 - - 1062 98

View Full Product Details